Läs mer
Trofinetide, MedChemExpress
Produktkod. 16433207
Encompass_Preferred
Change view
Kvantitet:
1 mg
5 mg
10 mg
50 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16433207 50 mg
16417176 100 mg
16409717 5 mg
16407188 10 mg
16401649 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16433207 Leverantör Medchem Express Leverantörsnummer HY16757
MedChemExpress Trofinetide (NNZ-2566), a synthetic analogue of the endogenous N-terminus tripeptide, Glycine-Proline-Glutamate (GPE), has been shown to be neuroprotective in animal models of brain injury.
Trofinetide (NNZ-2566), a synthetic analogue of the endogenous N-terminus tripeptide, Glycine-Proline-Glutamate (GPE), has been shown to be neuroprotective in animal models of brain injury.
Specifikationer
Kemiskt namn eller material Trofinetide
Fysisk form Powder
CAS 853400-76-7
Färg White
Kvantitet 50 mg
Anteckningar om renhetsgrad Research
Molekylformel C13H21N3O6
För användning med (applikation) Neuroscience-Neuromodulation
Synonym NNZ-2566
Löslighetsinformation H2O : ≥ 50 mg/mL (158.57 mM)
LEDER O=C(O)CC[C@@H](C(O)=O)NC([C@@]1(C)N(C(CN)=O)CCC1)=O
Molekylvikt (g/mol) 315.32
Formel vikt 315.32
Procent renhet 98.03%