Läs mer
Skytec™ Tromso+ Safety Gloves
Storlek:
Small
Medium
Large
X-Large
2X-Large
Beskrivning
- Thin and Flexible Fully Coated Insulated Glove with High Cut Protection
- Cut resistance : Level F / ANSI A6
- Fully dipped nitrile foam allowing good grip in wet and dry conditions
- Soft fleece lining for comfort in cold conditions
- Durable and lightweight coating ensures glove longevity in abrasive applications
- Good 360° breathability
- Ultra fine coating for maximum tactile sensitivity
- Touchscreen capabilities to allow usage with screen devices
- Minimized particle shedding
- Sanitised antimicrobial finish provides extra freshness
- Hand conforming fit for a second skin feel
- Contact heat protection up to 250°C / 482°F for a short period (15 sec. max.)
Specifikationer
Specifikationer
| Certifieringar/efterlevnad (Europa) | CE II, UKCA, ANSI/ISEA 105-2016: Cut A6,Puncture 5, EN 388:2016 + A1:2018 4X43F, EN 21420:2020 |
| Beläggningsmaterial | Nitrile |
| Färg | Black, Orange |
| Exteriör finish | Rough |
| Hand | Ambidextrous |
| Material | Foam Nitrile |
| Produkttyp | Cold-resistant Gloves |
| Storlek | Large |
| Temperatur (engelska) Max. | 482°F |
| Temperatur (metrisk) Max | 250°C |
| Visa mer |