Läs mer
Tropine, TRC
Produktkod. 30134832
Change view
Kvantitet:
10 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134832 100 g
30134809 50 g
17951185 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134832 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT892665100G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 120-29-6
Molekylformel C8 H15 N O
Molekylvikt (g/mol) 141.21
Synonym (1R,3r,5S)-8-Methyl-8-azabicyclo[3.2.1]oct-3-ol, 8-Azabicyclo[3.2.1]octan-3-ol, 8-methyl-, (3-endo)-, 1αH,5αH-Tropan-3α-ol (8CI), 8-Azabicyclo[3.2.1]octan-3-ol, 8-methyl-, endo-, Tropanol (6CI,7CI), (3-endo)-8-Methyl-8-azabicyclo[3.2.1]octan-3-ol, 2,3-Dihydro-3α-hydroxy-8-methylnortropidine, 2,3-Dihydro-3α-hydroxytropidine, 3α-Tropanol, NSC 43870, Tropine, α-Tropine
IUPAC-namn (1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-ol
LEDER CN1[C@@H]2CC[C@H]1C[C@@H](O)C2
Specifikationer
Kemiskt namn eller material Tropine
Kvantitet 100 g
InChI formel InChI=1S/C8H15NO/c1-9-6-2-3-7(9)5-8(10)4-6/h6-8,10H,2-5H2,1H3/t6-,7+,8+
Formel vikt 141.1154