missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Troponin I Type 1 (slow skeletal) Rabbit anti-Human, Rat, Clone: 5K1U2, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-16855-100UL
This item is not returnable.
View return policy
Description
Troponin I Type 1 (slow skeletal) Monoclonal antibody specifically detects Troponin I Type 1 (slow skeletal) in Human, Rat samples. It is validated for Western BlotSpecifications
Troponin I Type 1 (slow skeletal) | |
Monoclonal | |
Unconjugated | |
DKFZp451O223, slow-twitch isoform, SSTNI, TNN1, troponin I type 1 (skeletal, slow), troponin I, skeletal, slow, troponin I, slow skeletal muscle | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-187 of human Troponin I Type 1 (slow skeletal) (P19237). | |
100 μg | |
Cell Biology, Cellular Markers, Stem Cell Markers | |
7135 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
5K1U2 | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |