missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Troponin I Type 3 (cardiac) Antibody, Alexa Fluor™ 532, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP3-11029AF532
This item is not returnable.
View return policy
Description
Troponin I Type 3 (cardiac) Polyclonal antibody specifically detects Troponin I Type 3 (cardiac) in Human samples. It is validated for Western Blot, ELISASpecifications
Troponin I Type 3 (cardiac) | |
Polyclonal | |
Western Blot, ELISA | |
Cardiac troponin I, CMD1FF, CMD2A, CMH7MGC116817, cTnI, familial hypertrophic cardiomyopathy 7, RCM1, TNNC1troponin I, cardiac, troponin I type 3 (cardiac), troponin I, cardiac muscle | |
Purified Human Cardiac Troponin, I subunit | |
0.1 mL | |
Apoptosis, Cell Biology, Cellular Markers, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers | |
7137 | |
Store at 4°C in the dark. |
Western Blot, ELISA | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Goat | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |