Läs mer
Trospectomycin Dihydrochloride (>70%), TRC
Produktkod. 30452443
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30452443 1 mg
1 options
Produktkod. 30452443 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT8927501MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Trospectomycin Dihydrochloride
CAS 85951-37-7
Molekylformel C17H30N2O7• 2HCl
Kvantitet 1 mg
Synonym (2R,4aR,5aR,6S,7S,8R,9S,9aR,10aS)-2-Butyldecahydro-4a,7,9-trihydroxy-6,8-bis(methylamino)-4H-pyrano[2,3-b][1,4]benzodioxin-4-one Dihydrochloride
InChI formel InChI=1S/C17H30N2O7.2ClH/c1-4-5-6-8-7-9(20)17(23)16(24-8)25-15-13(22)10(18-2)12(21)11(19-3)14(15)26-17;;/h8,10-16,18-19,21-23H,4-7H2,1-3H3;2*1H/t8?,10-,11+,12+,13+,14-,15-,16+,17+;;/m1../s1
LEDER [H][C@]12[C@@](O)(O[C@@]3([C@H]([C@H]([C@H]([C@@H]([C@]3(O2)[H])O)NC)O)NC)[H])C(CC(O1)CCCC)=O.Cl.Cl
IUPAC-namn (4aR,5aR,6S,7S,8R,9S,9aR,10aS)-2-butyl-4a,7,9-trihydroxy-6,8-bis(methylamino)decahydro-4H-benzo[b]pyrano[2,3-e][1,4]dioxin-4-one dihydrochloride
Molekylvikt (g/mol) 447.35
Formel vikt 446.17