Läs mer
Troxerutin, MedChemExpress
Beskrivning
Troxerutin, also known as vitamin P4, is a tri-hydroxyethylated derivative of natural bioflavonoid rutins which can inhibit the production of reactive oxygen species (ROS) and depress ER stress-mediated NOD activation.
Specifikationer
Specifikationer
| Kemiskt namn eller material | Troxerutin |
| CAS | 7085-55-4 |
| Färg | Yellow |
| Anteckningar om renhetsgrad | Research |
| Molekylformel | C33H42O19 |
| Kvantitet | 100 mg |
| Synonym | Trihydroxyethylrutin |
| Löslighetsinformation | DMSO : 100 mg/mL (134.65 mM; Need ultrasonic and warming) |
| LEDER | O=C1C(O[C@H]2[C@@H]([C@H]([C@@H]([C@@H](CO[C@H]3[C@@H]([C@@H]([C@H]([C@H](C)O3)O)O)O)O2)O)O)O)=C(C4=CC=C(OCCO)C(OCCO)=C4)OC5=CC(OCCO)=CC(O)=C15 |
| Molekylvikt (g/mol) | 742.68 |
| Visa mer |