Läs mer
TRPV3 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29971-1-AP-20UL
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
The TRPV3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TRPV3. This antibody recognizes human antigen. The TRPV3 antibody has been validated for the following applications: IHC, ELISA analysis.Specifikationer
| TRPV3 | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 162514 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Immunohistochemistry, ELISA | |
| 750 μg/mL | |
| VRL3 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| Polyclonal Antibody | |
| IgG |