missing translation for 'onlineSavingsMsg'
Learn More

TRPV5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17322-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16827194

  • 1236.20 SEK / 20µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene is a member of the transient receptor family and the TrpV subfamily. The calcium-selective channel encoded by this gene has 6 transmembrane-spanning domains, multiple potential phosphorylation sites, an N-linked glycosylation site, and 5 ANK repeats. This protein forms homotetramers or heterotetramers and is activated by a low internal calcium level.
Specifications

Specifications

TRPV5
0.21 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P69744, Q9JIP0, Q9NQA5
Calcium transport protein 2, CAT2, ECaC, ECAC1, Epithelial calcium channel 1, Osm 9 like TRP channel 3, OTRPC3, TRPV5
Rabbit
IgG
20 μL
-20°C
Primary
116469, 194352, 56302
Immunocytochemistry, Immunofluorescence
Unconjugated
TRPV5
Liquid
TRPV5
TRPV5 Fusion Protein Ag11298
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Rat, Mouse
Videos
SDS
Documents
Certificates
Special Offers

Special Offers