missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRUB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19891-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Pseudouridine is an abundant component of rRNAs and tRNAs and is enzymatically generated by isomerization of uridine by pseudouridine synthase.Specifications
TRUB2 | |
Polyclonal | |
Unconjugated | |
TRUB2 | |
CLONE24922, RP11 339B21.1, TRUB2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
227682, 26995, 366012 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95900, Q5XFW2, Q91WG3 | |
Trub2 | |
TRUB2 Fusion Protein Ag13726 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |