Learn More
Lonza™ Trypsin Neutralizing Solution
Used to effectively neutralize Trypsin/EDTA. Lonza™ Trypsin Neutralizing Solution TNS is a component of the ReagentPack™ Subculture Reagent Kit
Brand: Lonza™ CC-5002
This item is not returnable.
View return policy
Description
- TLN
- A component of the ReagentPack™ Subculture Reagent Kit
- Solution 100 ml
Specifications
Trypsin inhibitor | |
100 mL |
Product Content Correction
Product Title
Lonza™ Trypsin Neutralizing Solution > Quantity: 100mL
Spot an opportunity for improvement?Share a Content Correction