Läs mer
Tryptanthrin, TRC
Produktkod. 30450393
Change view
Kvantitet:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30450393 5 mg
30454954 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30450393 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT9470005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tryptanthrin
CAS 13220-57-0
Molekylformel C15H8N2O2
Kvantitet 5 mg
Synonym Indolo[2,1-b]quinazoline-6,12-dione, Couroupitine A, NSC 349447, TryptanthrinTryptanthrine
InChI formel InChI=1S/C15H8N2O2/c18-13-10-6-2-4-8-12(10)17-14(13)16-11-7-3-1-5-9(11)15(17)19/h1-8H
LEDER O=C1N2C(=Nc3ccccc13)C(=O)c4ccccc24
IUPAC-namn indolo[2,1-b]quinazoline-6,12-dione
Molekylvikt (g/mol) 248.24
Formel vikt 248.0586