Läs mer
TSC22/TSC22D1 Antibody (040), Allophycocyanin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90142APC
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
TSC22/TSC22D1 Monoclonal antibody specifically detects TSC22/TSC22D1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifikationer
| Immunocytochemistry | |
| 040 | |
| PBS | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human TSC22/TSC22D1 (Uniprot#: Q15714-2; Met1-Ala144). | |
| Purified |
| Monoclonal | |
| APC | |
| Rabbit | |
| Store at 4C in the dark. |