missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSNAXIP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17730-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Possible role in spermatogenesis.Specifications
TSNAXIP1 | |
Polyclonal | |
Unconjugated | |
TSNAXIP1 | |
Trax interacting protein 1, TSNAXIP1, TXI1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55815, 72236 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.46 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q2TAA8, Q99P25 | |
TSNAXIP1 | |
TSNAXIP1 Fusion Protein Ag12130 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |