missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12812-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
TSPAN12 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility.The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility.Specifications
TSPAN12 | |
Polyclonal | |
Unconjugated | |
TSPAN12 | |
NET 2, NET2, Tetraspan NET 2, tetraspanin 12, TM4SF12, Tspan 12, TSPAN12 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23554, 269831, 362326 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95859, Q569A2, Q8BKT6 | |
Tspan12 | |
TSPAN12 Fusion Protein Ag3514 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |