missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSSK3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | TSSK3 |
---|---|
Concentration | 0.3 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890904
|
Proteintech
12940-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16880904
|
Proteintech
12940-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a kinase expressed exclusively in the testis that is thought to play a role in either germ cell differentiation or mature sperm function.Specifications
TSSK3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
Q96PN8, Q9D2E1 | |
297891, 58864, 81629 | |
TSSK3 Fusion Protein Ag4009 | |
Primary | |
-20°C | |
TSSK3 |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
SPOGA3, STK22C, STK22D, testis serine kinase 3, Testis specific kinase 3, TSK 3, TSSK 3, TSSK3 | |
Tssk3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |