missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSSK3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12940-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a kinase expressed exclusively in the testis that is thought to play a role in either germ cell differentiation or mature sperm function.Specifications
TSSK3 | |
Polyclonal | |
Unconjugated | |
TSSK3 | |
SPOGA3, STK22C, STK22D, testis serine kinase 3, Testis specific kinase 3, TSK 3, TSSK 3, TSSK3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
297891, 58864, 81629 | |
-20°C | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96PN8, Q9D2E1 | |
Tssk3 | |
TSSK3 Fusion Protein Ag4009 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |