missing translation for 'onlineSavingsMsg'
Learn More

TSTA3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15335-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16824474

  • 3313.85 SEK / 150µL
Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Tissue specific transplantation antigen P35B is a NADP(H)-binding protein. It catalyze the two-step epimerase and the reductase reactions in GDP-D-mannose metabolism, converting GDP-4-keto-6-D-deoxymannose to GDP-L-fucose. GDP-L-fucose is the substrate of several fucosyltransferases involved in the expression of many glycoconjugates, including blood group ABH antigens and developmental adhesion antigens. Mutations in this gene may cause leukocyte adhesion deficiency, type II.
Specifications

Specifications

TSTA3
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P23591, Q13630
FX, GDP L fucose synthase, P35B, Protein FX, SDR4E1, TSTA3
Rabbit
IgG
150 μL
-20°C
Primary
22122, 300036, 7264
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TSTA3
Liquid
TSTA3
TSTA3 Fusion Protein Ag7450
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers