Läs mer
Thermo Scientific™ TSU™ Series -86°C Upright Ultra-Low Temperature Freezers
Ensure ultimate protection and optimum capacity for your most critical samples with these upright ultra-low temperature freezers.
Beskrivning
Ensure ultimate protection and optimum capacity for your most critical samples with Thermo Scientific™ TSU™ Series −86°C Upright Ultra-Low Temperature Freezers. These freezers achieve outstanding thermal performance, safety and security through state-of-the-art engineering.
- Choice of five capacities maximize sample storage while minimizing the freezer's physical space inside the lab
- High-performance mode provides the tightest temperature uniformity and peak variation†—for most applications, energy-savings mode offers excellent temperature control, plus up to 15% savings on energy usage compared to high-performance mode
- Increases the internal capacity of 2 inch vials over previous generation freezers—gain up to 76% more capacity in the same footprint†— with 1 mL CryBank tubes
- Five sizes accommodate any size lab, featuring the industry's leading capacity per footprint specifications‡
- Monitor the freezer's health 24/7 and access a detailed event log
- Obtain a record, simply download a report of the current event log to a portable drive
- Environmentally-friendly, CFC/HCFC-free refrigerants
- High-efficiency compressors offer exceptional performance and reliability
- Brazed plate heat exchanger for more efficient heat transfer—induction brazed joints to reduce leak potential and improve reliability
- Power management system protects against a wide range of voltage variation and is easily accessible through the touch-screen display
- Remote alarm contacts compatible with external alarm and monitoring systems
- Two rear access, 1 in. (25 mm) ports allow for the use of inexpedient probes or instrumentation
- Store up to 15 years worth of temperature and event data on our on-board computer
- Use the new USB port to download freezer temperature and event log data, or freezer settings from one freezer to another
- Provides protection against dust on the condenser, which can cause reduced refrigeration performance and increased risk to samples
- Store up to 278 lbs. of samples (depending on freezer model) on our reinforced, stainless steel shelving
- 4×7 heated gasket provides four touchpoints of security and seven zones of protection, maximizing cabinet temperature and eliminating frost buildup
- Four polystyrene insulated inner doors help maintain cabinet temperature during openings and feature embedded rare earth magnets, eliminating exposed latches or magnets
- Several optional features including LN2 and CO2 backup system, chart recorder, stainless steel inventory racking solutions, and hands-free locking option
† Compared to energy-savings and high-performance modes.
‡ Based on internal performance data; Data on file May 2011
- Warranty varies by country, please contact us for details