Learn More
TTC26 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25083-1-AP-150UL
Additional Details : Weight : 0.00010kg
Specifications
TTC26 | |
Polyclonal | |
Unconjugated | |
TTC26 | |
dyf 13, TPR repeat protein 26, TTC26 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
264134, 79989 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A0AVF1, Q8BS45 | |
TTC26 | |
TTC26 Fusion Protein Ag18983 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |