missing translation for 'onlineSavingsMsg'
Learn More

TTC26 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 25083-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16823625

  • 1656.73 SEK / 20µL
Estimated Shipment: 25-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TTC26
Polyclonal
Unconjugated
TTC26
dyf 13, TPR repeat protein 26, TTC26
Rabbit
Antigen Affinity Chromatography
RUO
264134, 79989
-20°C
Liquid
Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
A0AVF1, Q8BS45
TTC26
TTC26 Fusion Protein Ag18983
20 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers