missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTLL10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | TTLL10 |
---|---|
Concentration | 0.23 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16824545
|
Proteintech
26014-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16814545
|
Proteintech
26014-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Inactive polyglycylase.Specifications
TTLL10 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
A4Q9F3, Q5XI57, Q6ZVT0 | |
254173, 298692, 330010 | |
TTLL10 Fusion Protein Ag23258 | |
Primary | |
-20°C | |
TTLL10 |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Inactive polyglycylase TTLL10, TTLL10, TTLL5 | |
TTLL10 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |