Läs mer
Tocris Bioscience™ TTNPB
GREENER_CHOICE
Produktkod. 30001123
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30001123 10 mg
18054512 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30001123 Leverantör Tocris Bioscience™ Leverantörsnummer 0761/10
Retinoic acid analog; RAR agonist
Extremely potent analog of retinoic acid, selective for the retinoic acid receptor (RAR) subtype. Enhances reprogramming efficiency in chemically induced pluripotent stem cells (CiPSCs).
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 71441-28-6
Molekylformel C24H28O2
Specifikationer
Kemiskt namn eller material TTNPB
Kvantitet 10 mg
Formel vikt 348.48
Renhet >98%
Molekylvikt (g/mol) 348.486