Learn More
Heidolph instruments Tube MonoBlock
Accommodate specifically sized reagent tubes at your StarFish magnetic stirrer workstation. Heidolph™ Tube MonoBlock, constructed of one piece, is ideal for experiments using the same vessel.
Brand:  Heidolph instruments 505-80005-00
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 11761699
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Accommodates one specific tube size.
  • One-piece construction.
Specifications
Specifications
MonoBlock
Starfish Workstation, MR-Hei Magnetic Stirrers
40 x 12 mm Tubes