Learn More
Heidolph instruments Tube MonoBlock
Accommodate specifically sized reagent tubes at your StarFish magnetic stirrer workstation. Heidolph™ Tube MonoBlock, constructed of one piece, is ideal for experiments using the same vessel.
Brand: Heidolph instruments 505-80005-00
Additional Details : Weight : 0.10000kg
Description
- Accommodates one specific tube size.
- One-piece construction.
Specifications
MonoBlock | |
Starfish Workstation, MR-Hei Magnetic Stirrers | |
40 x 12 mm Tubes |