missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ Tube rack, freezer, 7 x 4 holes 18mm diameter, polypropylene
Brand: Tenak™ TE50530
Additional Details : Weight : 0.55000kg
Specifications
Tube Rack | |
Tubes (18 mm) | |
28 | |
18 mm |