Läs mer
Tubocurarine Chloride, LoGiCal
Produktkod. 30460838
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30460838 5 mg
1 options
Produktkod. 30460838 Leverantör LoGiCal Leverantörsnummer LGCFOR1651.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material D-Tubocurarine Chloride
CAS 57-94-3
Analyt- eller komponentnamn D-Tubocurarine Chloride
Frakt skick Room Temperature
Molekylformel C37 H41 N2 O6 . Cl . H Cl
Certifikat och efterlevnad ISO 9001
Synonym (+)-Tubocurarine chloride hydrochloride, 2,3,13a,14,15,16,25,25a-Octahydro-9,19-dihydroxy-18,29-dimethoxy-1,14,14-trimethyl-13H-4,6:21,24-dietheno-8,12-metheno-1H-pyrido[3',2':14,15][1,11]dioxacycloeicosino[2,3,4-ij]isoquinolinium chloride hydrochloride
InChI formel InChI=1S/C37H40N2O6.2ClH/c1-38-14-12-24-19-32(42-4)33-21-27(24)28(38)16-22-6-9-26(10-7-22)44-37-35-25(20-34(43-5)36(37)41)13-15-39(2,3)29(35)17-23-8-11-30(40)31(18-23)45-33;;/h6-11,18-21,28-29H,12-17H2,1-5H3,(H-,40,41);2*1H/t28-,29+;;/m0../s1
Lösningstyp Neat
LEDER Cl.[Cl-].COc1cc2CC[N+](C)(C)[C@@H]3Cc4ccc(O)c(Oc5cc6[C@H](Cc7ccc(Oc(c1O)c23)cc7)N(C)CCc6cc5OC)c4
Industry Type Clinical Testing
Molekylvikt (g/mol) 681.645
Kvantitet 5 mg
Formel vikt 680.242 g/mol