Läs mer
Tucatinib, MedChemExpress
Produktkod. 16446978
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
100 mg
Förpackningsstorlek:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16446978 10 mg -
16417056 100 mg -
16413037 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16446978 Leverantör Medchem Express Leverantörsnummer HY16069
MedChemExpress Tucatinib (Irbinitinib) is a potent, orally active and selective HER2 inhibitor with an IC50 of 8 nM.
Tucatinib (Irbinitinib) is a potent, orally active and selective HER2 inhibitor with an IC50 of 8 nM.
Specifikationer
Kemiskt namn eller material Tucatinib
CAS 937263-43-9
Färg Light Yellow
Anteckningar om renhetsgrad Research
Molekylformel C26H24N8O2
Kvantitet 10 mg
Synonym Irbinitinib ARRY-380 ONT-380
Löslighetsinformation DMSO : 41.67 mg/mL (86.72 mM; ultrasonic and warming and heat to 60°C)
LEDER CC1=CC(NC2=C3C=C(NC4=NC(C)(C)CO4)C=CC3=NC=N2)=CC=C1OC5=CC6=NC=NN6C=C5
Molekylvikt (g/mol) 480.52
Formel vikt 480.52
Procent renhet 98.47%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease