Läs mer
Thermo Scientific™ Tulobuterol
beta-2-adrenergic receptor agonist
Brand: Thermo Scientific™ J63975.03
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifikationer
| 1 g | |
| -30°C to -10°C | |
| beta-2-adrenergic receptor agonist | |
| C12H18ClNO | |
| 159°C to 163°C |
| White to Off-white | |
| Crystalline Powder | |
| 41570-61-0 | |
| 227.73g/mol |