Läs mer
Tungstosilicic Acid Hydrate, TRC
Produktkod. 30455275
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30455275 50 mg
30451940 100 mg
30454774 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30455275 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT89727350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 12027-38-2
Molekylformel H4[Si(W3O10)4] . xH2O
Molekylvikt (g/mol) 2896.18
Synonym Tungstosilicic Acid (H4SiW12O40) (7CI,8CI), Silicic Acid (H4SiO4), Tungsten Complex, 12-Tungstosilicic Acid, 12-Tungstosilicic Acid (H4W12O40Si), Dodecatungstosilicic Acid (H4SiW12O40), JM 1493, NSW 40, PM 92, Silicotungstic Acid, Tungsten Hydroxide Oxide Silicate, (W12(OH)4O32(SiO4)), Tungsten Oxide Silicide (W12O38Si), Tungstosilicic Acid (H4W12SiO40)
IUPAC-namn silicic acid;trioxotungsten
LEDER O[Si](O)(O)O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O
Specifikationer
Kemiskt namn eller material Tungstosilicic Acid Hydrate
Kvantitet 50 mg
InChI formel InChI=1S/H4O4Si.36O.12W/c1-5(2,3)4;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/h1-4H;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;
Formel vikt 2879.216