missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
STEMCELL Technologies TWS119
Beskrivning
- TWS119 is a potent disubstituted pyrrolopyrimidine inhibitor of glycogen synthase kinase 3 beta (GSK3β) with an IC50 of 30 nM and Kd of 126 nM (Ding et al.) GSK3 is a serine/threonine kinase that is a key inhibitor of the WNT pathway; therefore TWS119 functions as a WNT pathway activator
- Maintains the bi-potent, quiescent state in hepatic stellate cells of Wistar rats (Kordes et al.)
- Induces differentiation of mouse embryonal carcinoma and embryonic stem cells to neurons (Ding et al.)
- Induces production of T memory stem cell-like (T-SCM) cells from mouse or human CD8+ T cells with evidence of increased persistence, proliferation, and anti-tumor activity after adoptive transfer of mouse-derived T-SCM (Forget et al.; Gattinoni et al.)
- Inhibits cell proliferation and induces apoptosis in human alveolar rhabdomyosarcoma cells (Zeng et al.)
Specifikationer
Specifikationer
| CAS | 601514-19-6 |
| Synonym | GSK 3B Inhibitor XII |
| Kvantitet | 10 mg |
| Molekylformel | C18H14N4O2 |
| Renhet | ≥90% |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?