Learn More
Thermo Scientific™ TX-750 Round Bucket Adapters
Add functionality to your rotor with round bucket adapters, which provide support for round-bottom and conical tubes and bottles, microtubes, and blood collection tubes.
Brand:  Thermo Scientific™ 75003719
8678.86 SEK valid until 2025-07-04
Use promo code "24147" to get your promotional price.
 View more versions of this product
Product Code. 11874461
  • 9135.64 SEK / Pack of 4
Quality Analysis and Quality Control Testing
Use promo code 24140 at checkout
View Available Promotions
Description
Add functionality to your rotor with Thermo Scientific™ TX-750 Round Bucket Adapters, which provide support for round-bottom and conical tubes and bottles, microtubes and blood collection tubes.
Adapters will hold round-bottom and conical tubes and bottles, microtubes and blood collection tubes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Round Bucket Adapter
15 mL Blood Collection Tube
84 x 15 mL Blood Collection Tube
Round-Bottom and Conical Tube and Bottle, Microtube and Blood Collection Tube
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ TX-750 Round Bucket Adapters > Capacity: 84 x 15mL