Läs mer
Eurofins DiscoverX Txk Protein, Active
Beskrivning
TXK (TXK Tyrosine Kinase) is a ,on-receptor tyrosine kinase that plays a redundant role with ITK in regulation of the adaptive immune response. It predicted to enable non-membrane spanning protein tyrosine kinase activity. TXK is also involved in positive regulation of interferon-gamma-mediated signaling pathway, positive regulation of macromolecule metabolic process, and protein autophosphorylation. TXK is located in cytoplasm and nucleus.
GenBank NM_003328
Specifikationer
Specifikationer
| Formulering | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1 mM EGTA, 0.1% 2-mercaptoethanol, 0.03% Brij-35. Frozen solution. |
| Molekylvikt (g/mol) | 35.3 kDa |
| Kvantitet | 250 μg |
| Art | Human |
| Rekombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Uttryckssystem | Recombinant enzyme expressed in Sf21 insect cells |
| Proteinform | Truncated |
| Renhet eller kvalitetsklass | ≥51% |
| Protein | Txk, active |
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.