missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXK Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18981-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The Tec family of non-receptor tyrosine kinases is composed of six proteins designated Tec, Emt (also known as Itk or Tsk), Btk (previously known as Atk, BPK or Emb), Bmx, Txk (also known as Rlk) and Dsrc28C. All members of the family contain SH3 and SH2 domains and, with the exception of Txk and Dsrc28C, also contain a pleckstrin homology (PH) and a Tec homology (TH) domain in their amino termini. Four alternatively spliced forms of Tec are found to be expressed broadly in cells of hematopoietic lineage and hepatocytes. The Emt gene product associates with CD28 and becomes activated subsequent to CD28 ligation. Btk is necessary for proper B cell development, and mutations in the gene encoding Btk have been associated with families suffering from X-linked agammaglobulinemia, also referred to as Bruton′s disease. The Bmx protein shares a high degree of homology with Btk and seems to be expressed at highest levels in the heart. Txk expression is T cell-specific, while expression of the Drosophila Tec homolog, Dsrc28C, is developmentally regulated.Specifications
TXK | |
Polyclonal | |
Unconjugated | |
TXK | |
BTKL, Protein tyrosine kinase 4, PSCTK5, PTK4, RLK, TKL, TXK, TXK tyrosine kinase, Tyrosine protein kinase TXK | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7294 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P42681 | |
TXK | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |