missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TXK, GST Tag Recombinant Protein
TXK Recombinant Human Protein
5330.00 SEK - 18240.00 SEK
Specifications
Accession Number | NP_003319.2 |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 58.5 kDa |
Gene Alias | TXK |
KinaseFamily | TEC Kinase Family |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10277564
|
Invitrogen
PV5860 |
10μg |
5330.00 SEK
Each |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15438713
|
Invitrogen
PR9089A |
100μg |
18240.00 SEK
100µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
TXK, along with the close relatives BMS, ITK, and BTK, signal downstream of antigen receptors and other types of receptors.Specifications
NP_003319.2 | |
58.5 kDa | |
TEC Kinase Family | |
Recombinant | |
Kinase proteins are shipped on dry ice and should be stored at -80°C. | |
Dry Ice | |
Kinases & Inhibitors | |
Liquid |
Kinase Assay | |
TXK | |
Human | |
GST Tag | |
Tyrosine Kinases | |
Baculovirus | |
Recombinant, Catalytic Domain |
For Research Use Only. Not for use in diagnostic procedures.