missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNDC17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20811-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
TXNL5 disulfide reductase. May participate in various redox reactions through the reversible oxidation of its active center dithiol to a disulfide and catalyze dithiol-disulfide exchange reactions. Modulates TNF-alpha signaling and NF-kappa-B activation. Has peroxidase activity and may contribute to the elimination of cellular hydrogen peroxide (By similarity).Specifications
TXNDC17 | |
Polyclonal | |
Unconjugated | |
TXNDC17 | |
Protein 42 9 9, Thioredoxin like protein 5, TRP14, TXNDC17, TXNL5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
287474, 52700, 84817 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BRA2, Q9CQM5 | |
Txndc17 | |
TXNDC17 Fusion Protein Ag14766 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |