missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ TXNL4A Recombinant Protein

Brand:  Abnova™ H00010907-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16135832

  • 4515.00 SEK / 10µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH01046
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
41.36
8
Glutathione Sepharose 4 Fast Flow
10 ug
MSYMLPHLHNGWQVDQAILSEEDRVVVIRFGHDWDPTCMKMDEVLYSIAEKVKNFAVIYLVDITEVPDFNKMYELYDPCTVMFFFRNKHIMIDLGTGNNNKINWAMEDKQEMVDIIETVYRGARKGRGLVVSPKDYSTKYRY
DIB1/DIM1/HsT161/TXNL4/U5-15kD
TXNL4A
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
10907
TXNL4A (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TXNL4A
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers