missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNL6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11203-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Retinitis pigmentosa (RP) is a disease that leads to blindness by degeneration of cone photoreceptors. Rods produce factors required for cone viability. The protein encoded by this gene is one of those factors and is similar to a truncated form of thioredoxin. This gene has been proposed to have therapeutic value against RP.Specifications
TXNL6 | |
Polyclonal | |
Unconjugated | |
NXNL1 | |
nucleoredoxin like 1, Nucleoredoxin like protein 1, NXNL1, RDCVF, Thioredoxin like protein 6, TXNL6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
115861, 234404 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8VC33, Q96CM4 | |
Nxnl1 | |
TXNL6 Fusion Protein Ag1691 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |