Learn More
Ismatec™ Tygon™ E-LFL Microbore 2-Stop Tube Sets
Longest life of all Tygon™ formulations
Brand:  Ismatec™ 06447-42
Additional Details : Weight : 0.04000kg
 View more versions of this product
Product Code. 15560652
  • 961.00 SEK / Pack of 12
Estimated Shipment
02-02-2021
Add to basket
Description
Description
  • Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • For use with Masterflex™ L/S™ multichannel pump heads
Specifications
Specifications
USP; EU Food 10/2011; European Pharmacopoeia; Non-DEHP; Non-Phthalate; RoHS; Prop 65 NA (no Prop 65 chemicals)
2.06 mm
1.25 ft.
L/S Series Stopped Pump Tubing
Pk. of 12
130 mL/min
25.3 psi
Tygon E-LFL
0.38 m
0°C
42
3/32 in.