Learn More
TYK2 Mouse anti-Human, Unconjugated, Clone: 1E1A10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67411-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The TYK2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TYK2. This antibody recognizes human antigen. The TYK2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
TYK2 | |
Monoclonal | |
800 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, ELISA | |
1E1A10 | |
Unconjugated | |
20 μL | |
7297 | |
IgG2b |