Läs mer
Tymazoline Hydrochloride, TRC
Produktkod. 30454834
Change view
Kvantitet:
250 mg
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30454834 2.5 g 2.5g
30449378 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30454834 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT8982502.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 28120-03-8
Molekylformel C14 H20 N2 O . Cl H
Molekylvikt (g/mol) 268.78
Synonym 1H-Imidazole, 4,5-dihydro-2-[[5-methyl-2-(1-methylethyl)phenoxy]methyl]-, hydrochloride (1:1), 1H-Imidazole, 4,5-dihydro-2-[[5-methyl-2-(1-methylethyl)phenoxy]methyl]-, monohydrochloride (9CI), 2-Imidazoline, 2-[(thymyloxy)methyl]-, monohydrochloride (8CI), 2-(Thymyloxymethyl)glyoxalidine hydrochloride, Thymazen, Tymazoline hydrochloride
IUPAC-namn 2-[(5-methyl-2-propan-2-ylphenoxy)methyl]-4,5-dihydro-1H-imidazole;hydrochloride
LEDER Cl.CC(C)c1ccc(C)cc1OCC2=NCCN2
Specifikationer
Kemiskt namn eller material Tymazoline Hydrochloride
Kvantitet 2.5 g
InChI formel InChI=1S/C14H20N2O.ClH/c1-10(2)12-5-4-11(3)8-13(12)17-9-14-15-6-7-16-14;/h4-5,8,10H,6-7,9H2,1-3H3,(H,15,16);1H
Formel vikt 268.1342