missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Cabinet Q-CLASSIC-90 Self Closing

Safe and approved storage of flammable hazardous substances in working areas

Brand:  asecos™ 34619-040-34617

Additional Details : Weight : 306.25201kg

 View more versions of this product

Product Code. 15967666

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.

Alert:

For indoor use only


Description

Description

  • Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
  • Easy handling with comfort: door stops in any position, self-closing in the event of fire
  • Easy handling with comfort: smooth doors with permanent self-closing function and oil-dampened door closer; open doors with a minimum amout of force
  • No unauthorised use: door lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
  • Easy transport: integrated transport base with removable, optional base cover
  • Easy alignment: adjusting aids to compensate for uneven floor
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system, spinning wheel in the exhaust grid as an indicator of sufficient technical ventilation
  • Triple certified: cabinet tested in accordance with EN 14470-1, FM Standard 6050 and UL/ULC Standard 1275
Specifications

Specifications

1647 mm
Safety storage cabinet
450 mm
Powder-coated textured
599 mm
315 kg
Anthracite Gray, Warning Yellow
615 mm
Wing, Right-hinged, Self-closing
3
1
3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated)
Safe and approved storage of flammable hazardous substances in working areas
522 mm
Steel
615 x 599 x 1953 mm
522 x 450 x 1647 mm
Standard
1953 mm
Q90.195.060.FU.WDCR
90 mm
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant, FM 6050 approved, UL/ULC 1275 listed
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers