missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet Q-CLASSIC-90 Self Closing
Safe and approved storage of flammable hazardous substances in working areas
22690.00 SEK - 29247.08 SEK
Specifications
Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant, FM 6050 approved, UL/ULC 1275 listed |
---|---|
Configuration | 3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) |
Depth (Metric) Exterior | 615 mm |
Depth (Metric) Interior | 522 mm |
Material | Steel |
Alert:
For indoor use only
Product Code | Brand | Width (Metric) Exterior | Door Style | Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) Exterior | Door Style | Color | Price | Quantity & Availability | |||
15907676
|
asecos™
34619-041-34617 |
599 mm | Right-hinged | Anthracite Gray, Light Gray |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15927666
|
asecos™
34610-041-34617 |
599 mm | Wing, Left-hinged, Self-closing | Anthracite Gray, Light Gray |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15927676
|
asecos™
34623-040-34628 |
893 mm | Wing, Self-closing | Anthracite Gray, Warming Yellow |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15937686
|
asecos™
34632-041-34637 |
1193 mm | Wing, Self-closing | Anthracite Gray, Light Gray |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15967666
|
asecos™
34619-040-34617 |
599 mm | Wing, Right-hinged, Self-closing | Anthracite Gray, Warning Yellow |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15967676
|
asecos™
34623-041-34628 |
893 mm | Wing, Self-closing | Anthracite Gray, Light Gray |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15987656
|
asecos™
34610-040-34617 |
599 mm | Wing, Left-hinged, Self-closing | Anthracite Gray, Warning Yellow |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
- Easy handling with comfort: door stops in any position, self-closing in the event of fire
- Easy handling with comfort: smooth doors with permanent self-closing function and oil-dampened door closer; open doors with a minimum amout of force
- No unauthorised use: door lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
- Easy transport: integrated transport base with removable, optional base cover
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system, spinning wheel in the exhaust grid as an indicator of sufficient technical ventilation
- Triple certified: cabinet tested in accordance with EN 14470-1, FM Standard 6050 and UL/ULC Standard 1275
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant, FM 6050 approved, UL/ULC 1275 listed | |
615 mm | |
Steel | |
Safe and approved storage of flammable hazardous substances in working areas | |
1647 mm | |
Standard |
3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | |
522 mm | |
Safety storage cabinet | |
1953 mm | |
3 | |
Powder-coated textured |