Learn More
Macherey-Nagel™ Type N24 Clear Screw Neck Vials
Brand:  Macherey-Nagel™ 702023
Product Code. 15236288
  • 510.00 SEK / Pack of 100
In Stock
to see stock.
Explore more special offers
Specifications
Clear
40 mL
With Closed Top Screw
100
Flat
100 Pk.
27.5 mm
For VOC / TOC analyses
Screw Neck, With Closed Top Screw
100 Pack
95 mm
Screw Thread Vial