Läs mer
Thermo Scientific™ [Tyr6, D-Phe7, D-His9] Substance P (6-11)
Kvantitet:
5 mg
10 mg
Förpackningsstorlek:
10mg
5mg
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifikationer
Specifikationer
| Reningsmetod | HPLC |
| Vanligt namn | Sendide |
| Innehåll och lagring | -30°C to -10°C |
| Formatera | Solid |
| Molekylvikt (g/mol) | 856.05 |
| Produkttyp | [Tyr6, D-Phe7, D-His9] Substance P (6-11) |
| Kvantitet | 5 mg |
| Regulatorisk status | RUO - research use only |
| Sekvens | Tyr-D-Phe-Phe-D-His-Leu-Met- NH2 |
For Research Use Only