Learn More
DuPont™ Tyvek™ 500 Laboratory Coats Press Studs
Offers enhanced protection for body parts that are more exposed to hazardous substances, fine particles, and fibres. Other features include ultra low-linting and antistatically treated.
463.75 SEK - 4265.00 SEK
Specifications
Product Type | Chemical Resistant Lab Coat |
---|---|
Gender | Unisex |
Material | HDPE |
Closure Type | Press Studs |
Disposable | Limited-use |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
11742594
|
DuPont™
D13396069 |
Large |
4265.00 SEK
Pack of 50 |
Estimated Shipment: 22-07-2025
Log in to see stock. |
||||||
15541362
|
DuPont™
D13396079 |
Medium | N/A | |||||||
15531362
|
DuPont™
D13396050 |
X-Large | N/A | |||||||
11752594
|
DuPont™
D13396040 |
2X-Large |
4265.00 SEK
Pack of 50 |
Estimated Shipment: 25-07-2025
Log in to see stock. |
||||||
Description
- Silicon non-added
- Stitched internal seams
- 3 pockets and 5 press stud closures
- Composed of flash spun high density polyethylene
- Provides an idea balance of protection, durability and comfort
- Permeable to both air and water vapor, yet repels water-based liquids and aerosols
- Applications include pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up
Specifications
Chemical Resistant Lab Coat | |
HDPE | |
Limited-use | |
Chemicals |
Unisex | |
Press Studs | |
Non-sterile |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
DuPont™ Tyvek™ 500 Laboratory Coats Press Studs