Learn More
Eppendorf™ U570 Inner Shelf K.O.P.
Brand:  Eppendorf™ K0280-1030
Product Code. 11654311
  • 1485.00 SEK / Each
Estimated Shipment: 25-09-2024
to see stock.
Explore more special offers
Description
Freezer Extra Shelf for CryoCube™ F570/ F570/ F570hw, includes four mounting clips, U570
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Freezers