missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Ub-K33 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120623
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Positive test controls include: HeLa, NIH/3T3, RAW264.7, C6.
Exists either covalently attached to another protein, or free (unanchored). When covalently bound, it is conjugated to target proteins via an isopeptide bond either as a monomer (monoubiquitin), a polymer linked via different Lys residues of the ubiquitin (polyubiquitin chains) or a linear polymer linked via the initiator Met of the ubiquitin (linear polyubiquitin chains). Polyubiquitin chains, when attached to a target protein, have different functions depending on the Lys residue of the ubiquitin that is linked: Lys-6-linked may be involved in DNA repair; Lys-11-linked is involved in ERAD (endoplasmic reticulum-associated degradation) and in cell-cycle regulation; Lys-29-linked is involved in lysosomal degradation; Lys-33-linked is involved in kinase modification; Lys-48-linked is involved in protein degradation via the proteasome; Lys-63-linked is involved in endocytosis, DNA-damage responses as well as in signaling processes leading to activation of the transcription factor NF-kappa-B. Linear polymer chains formed via attachment by the initiator Met lead to cell signaling. Ubiquitin is usually conjugated to Lys residues of target proteins, however, in rare cases, conjugation to Cys or Ser residues has been observed. When polyubiquitin is free (unanchored-polyubiquitin), it also has distinct roles, such as in activation of protein kinases, and in signaling. [Uniprot].
Specifikationer
| Ub-K33 | |
| Polyclonal | |
| Unconjugated | |
| UBB | |
| UBB | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Dot Blot, ELISA, Western Blot | |
| 0.68 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P0CG47, P0CG49, P0CG51 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 192255, 22187, 7314 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering