Learn More
UBC12 Mouse anti-Human, Unconjugated, Clone: 1H8D2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67482-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16840136
  • 1236.20 SEK / 20µL
Estimated Shipment: 05-10-2022
to see stock.
Description
Description
The UBC12 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human UBC12. This antibody recognizes human, mouse, rat antigen. The UBC12 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
UBC12
Monoclonal
1000 μg/mL
Mouse
Primary
Mouse, Rat, Human
ELISA, Immunohistochemistry, Western Blot
1H8D2
Unconjugated
20 μL
9040
IgG1