missing translation for 'onlineSavingsMsg'
Learn More

UBC9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 51018-2-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16826285

  • 1211.96 SEK / 20µL
Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

In heart and muscle extracts the UBE2I antibodies recognizes a 38 kDa protein band, but this band is not visible in extracts of other rat tissues.

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. Four alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications

Specifications

UBC9
0.6 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P63279, P63280
C358B7.1, E2,UBE2I, P18, SUMO conjugating enzyme UBC9, SUMO protein ligase, UBC9, UBCE9, UBE2I, Ubiquitin carrier protein 9, Ubiquitin carrier protein I, Ubiquitin protein ligase I
Rabbit
IgG
20 μL
-20°C
Primary
22196, 7329
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
UBE2I
Liquid
UBE2I
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers