Learn More
UBE1 Mouse anti-Human, Unconjugated, Clone: 1H7E8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67198-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The UBE1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human UBE1. This antibody recognizes human, mouse, rat antigen. The UBE1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
UBE1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, Immunofluorescence | |
1H7E8 | |
Unconjugated | |
150 μL | |
7317 | |
IgG2a |